This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SPI1 SIRNA Assays ›
Gene Symbol:
Spi1
Gene Name:
Spi-1 proto-oncogene
Gene Aliases:
Pu.1, Sfpi1
Chromosome Location:
Chr.3: 79918135 - 79937708 on Build Rnor_6.0
UniGene ID:
Rn.230674
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s172582

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Spi1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005892.2 NP_001005892.1 5 663
XM_006234532.2 XP_006234594.1 5 1542
GenBank mRNA AB154364.1 - 5 526
BC091185.1 - 5 663 7366435

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
vasculature development
lymphoid progenitor cell differentiation
myeloid leukocyte differentiation
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
lymphocyte differentiation
cell differentiation
erythrocyte differentiation
macrophage differentiation
granulocyte differentiation
somatic stem cell population maintenance
myeloid dendritic cell differentiation
histone H3 acetylation
hypermethylation of CpG island
negative regulation of MHC class II biosynthetic process
regulation of erythrocyte differentiation
negative regulation of gene expression, epigenetic
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
anatomical structure regression
cellular response to ethanol
negative regulation of histone H4 acetylation
apoptotic process involved in patterning of blood vessels
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II core promoter sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II core promoter proximal region sequence-specific binding
core promoter binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II transcription factor binding
transcriptional activator activity, RNA polymerase II distal enhancer sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
protein binding
sequence-specific DNA binding
NFAT protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0