This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CYB5R2 SIRNA Assays ›
Gene Symbol:
Cyb5r2
Gene Name:
cytochrome b5 reductase 2
Gene Aliases:
RGD1308421
Chromosome Location:
Chr.1: 171971424 - 171980641 on Build Rnor_6.0
UniGene ID:
Rn.16198
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s172046

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cyb5r2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014244.1 NP_001014266.1 5 486
XM_008759774.1 XP_008757996.1 3 1624
XM_008759775.1 XP_008757997.1 4 1527
XM_008759776.1 XP_008757998.1 3 1624
XM_008759777.1 XP_008757999.1 3 1624
XM_008759778.1 XP_008758000.1 4 942
XM_008759779.1 XP_008758001.1 4 1099
XM_008759780.1 XP_008758002.1 2 128
XM_008759781.1 XP_008758003.1 5 423
XM_008759782.1 XP_008758004.1 3 1624
GenBank mRNA BC079235.1 - 5 486 7132617

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

sterol biosynthetic process
oxidation-reduction process
cytochrome-b5 reductase activity, acting on NAD(P)H

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header