siRNA Details
Gene Symbol: Thsd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108878.1 | NP_001102348.1 | 3 | 591 | ||
XM_006253357.2 | XP_006253419.1 | 3 | 2009 | |||
GenBank mRNA | BC162035.1 | - | 2 | 462 | 7097232 |
