This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KCTD9 SIRNA Assays ›
Gene Symbol:
Kctd9
Gene Name:
potassium channel tetramerization domain containing 9
Gene Aliases:
-
Chromosome Location:
Chr.15: 44411378 - 44439645 on Build Rnor_6.0
UniGene ID:
Rn.9317
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s171461

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Kctd9
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108871.1 NP_001102341.1 1, 2 131
XM_006252138.2 XP_006252200.1 1, 2 117
XM_006252139.2 XP_006252201.1 1, 2 236
GenBank mRNA FQ221845.1 - 1, 2 241

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001285933, NM_134073, NM_001111028, AK166501, AK158922, and others.

Gene Ontology Categories:

Function(s)

protein homooligomerization

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0