siRNA Details
Gene Symbol: RGD1561958 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108819.1 | NP_001102289.1 | 6 | 736 | ||
XM_006257192.2 | XP_006257254.1 | 6 | 683 |

Gene Symbol: RGD1561958 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108819.1 | NP_001102289.1 | 6 | 736 | ||
XM_006257192.2 | XP_006257254.1 | 6 | 683 |