This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CAV2 SIRNA Assays ›
Gene Symbol:
Cav2
Gene Name:
caveolin 2
Gene Aliases:
-
Chromosome Location:
Chr.4: 44573264 - 44580640 on Build Rnor_6.0
UniGene ID:
Rn.81070
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s170732

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cav2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_131914.2 NP_571989.2 1 224
GenBank mRNA AF439780.1 - 1 101
BC062059.1 - 1 224 5598269
FQ209775.1 - 1 278

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of endothelial cell proliferation
positive regulation of endothelial cell proliferation
vesicle fusion
mitochondrion organization
endoplasmic reticulum organization
regulation of mitotic nuclear division
chemical synaptic transmission
negative regulation of cell proliferation
insulin receptor signaling pathway
vesicle organization
negative regulation of transforming growth factor beta receptor signaling pathway
positive regulation of MAPK cascade
positive regulation of GTPase activity
vesicle docking
skeletal muscle fiber development
positive regulation of peptidyl-tyrosine phosphorylation
protein oligomerization
positive regulation of dopamine receptor signaling pathway
caveola assembly
positive regulation of protein localization to nucleus
SNARE binding
protein binding
protein kinase binding
syntaxin binding
protein binding, bridging
mitogen-activated protein kinase kinase kinase binding
D1 dopamine receptor binding
protein complex scaffold
protein homodimerization activity
protein heterodimerization activity
phosphoprotein binding
scaffold protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0