This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PDE6D SIRNA Assays ›
Gene Symbol:
Pde6d
Gene Name:
phosphodiesterase 6D, cGMP-specific, rod, delta
Gene Aliases:
-
Chromosome Location:
Chr.9: 93562098 - 93607076 on Build Rnor_6.0
UniGene ID:
Rn.44816
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s170651

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pde6d
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108806.1 NP_001102276.1 2 204
XM_006245413.2 XP_006245475.1 2 252

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008801, BC005636, AK165701, AK170067, AF039216, and others.

Gene Ontology Categories:

Function(s) Process(es)

visual perception
metabolic process
negative regulation of catalytic activity
3',5'-cyclic-nucleotide phosphodiesterase activity
GTPase inhibitor activity
Rab GTPase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0