This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PPP2R5D SIRNA Assays ›
Gene Symbol:
Ppp2r5d
Gene Name:
protein phosphatase 2, regulatory subunit B', delta
Gene Aliases:
-
Chromosome Location:
Chr.9: 16580985 - 16610429 on Build Rnor_6.0
UniGene ID:
Rn.155910
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s170517

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ppp2r5d
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001062510.5 XP_001062510.4 14 2214
XM_006244570.2 XP_006244632.1 14 2217
GenBank mRNA BC099800.1 - 13 1506

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0