This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LOC100909464 SIRNA Assays ›
Gene Symbol:
LOC100909464 Ppp2r5d
Gene Name:
serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit delta isoform-like
protein phosphatase 2, regulatory subunit B', delta
Gene Aliases:
-
-
Chromosome Location:
Chr.9: 603900 - 606053 on Build Rnor_6.0
Chromosome Location:
Chr.9: 16580985 - 16610429 on Build Rnor_6.0
UniGene ID:
-
Rn.155910
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID s170516

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LOC100909464
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003750611.3 XP_003750659.3 1 214
Gene Symbol: Ppp2r5d
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001062510.5 XP_001062510.4 4 1080
XM_006244570.2 XP_006244632.1 4 1083
GenBank mRNA BC099800.1 - 3 372

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

signal transduction
regulation of catalytic activity
protein phosphatase type 2A regulator activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0