This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RANGAP1 SIRNA Assays ›
Gene Symbol:
Rangap1
Gene Name:
RAN GTPase activating protein 1
Gene Aliases:
-
Chromosome Location:
Chr.7: 122939263 - 122967178 on Build Rnor_6.0
UniGene ID:
Rn.107099
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s170109

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rangap1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001244855.1 NP_001231784.1 2, 3 336
XM_006242100.2 XP_006242162.1 2, 3 1776
XM_006242101.2 XP_006242163.1 2, 3 625
GenBank mRNA BC082056.1 - 2, 3 314 7132303
CV126932.1 - 2, 3 336
FQ217036.1 - 2, 3 320

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011241, NM_001146174, BC014855, AK133905, AK145459, and others.

Gene Ontology Categories:

Function(s)

signal transduction
negative regulation of protein export from nucleus

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0