This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TMEM184B SIRNA Assays ›
Gene Symbol:
Tmem184b
Gene Name:
transmembrane protein 184B
Gene Aliases:
RGD1306591
Chromosome Location:
Chr.7: 120593336 - 120636599 on Build Rnor_6.0
UniGene ID:
Rn.23598
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s170098

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tmem184b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001173370.1 NP_001166841.1 3 471
XM_006242013.2 XP_006242075.1 3 478
XM_006242014.2 XP_006242076.1 3 1064
GenBank mRNA BC099785.1 - 3 471

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0