This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other UQCRB SIRNA Assays ›
Gene Symbol:
Uqcrb
Gene Name:
ubiquinol-cytochrome c reductase binding protein
Gene Aliases:
Uqcrbl
Chromosome Location:
Chr.7: 71264501 - 71269869 on Build Rnor_6.0
UniGene ID:
Rn.105517
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s169990

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Uqcrb
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127553.2 NP_001121025.1 3 236
GenBank mRNA BC166882.1 - 3 221 6919310
CB783276.1 - 3 236
FQ217398.1 - 3 221
FQ217747.1 - 1 32
FQ217799.1 - 3 235

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

mitochondrial electron transport, ubiquinol to cytochrome c
protein heterooligomerization
protein complex binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0