siRNA Details
Gene Symbol: Uqcrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127553.2 | NP_001121025.1 | 4 | 880 | ||
GenBank mRNA | AW918593.1 | - | 1 | 492 |

Gene Symbol: Uqcrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127553.2 | NP_001121025.1 | 4 | 880 | ||
GenBank mRNA | AW918593.1 | - | 1 | 492 |