This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other HSP90B1 SIRNA Assays ›
Gene Symbol:
Hsp90b1
Gene Name:
heat shock protein 90, beta, member 1
Gene Aliases:
Grp94, Tra1
Chromosome Location:
Chr.7: 27226570 - 27240533 on Build Rnor_6.0
UniGene ID:
Rn.101146
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s169929

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Hsp90b1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012197.2 NP_001012197.2 12 1685
GenBank mRNA BC081917.1 - 12 1685 7112411
FQ229309.1 - 1 99

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011631, BC011439, AK209332, AK165975, AK031918, and others.

Gene Ontology Categories:

Function(s) Process(es)

response to hypoxia
protein folding
ER-associated ubiquitin-dependent protein catabolic process
actin rod assembly
negative regulation of apoptotic process
regulation of phosphoprotein phosphatase activity
cellular response to ATP
RNA binding
ATP binding
protein phosphatase binding
virion binding
low-density lipoprotein particle receptor binding
unfolded protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0