This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MBIP SIRNA Assays ›
Gene Symbol:
Mbip
Gene Name:
MAP3K12 binding inhibitory protein 1
Gene Aliases:
-
Chromosome Location:
Chr.6: 77226439 - 77243695 on Build Rnor_6.0
UniGene ID:
Rn.24202
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s169679

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mbip
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108712.2 NP_001102182.2 3, 4 543
XM_006240120.2 XP_006240182.1 3, 4 561
GenBank mRNA BC166982.1 - 3, 4 470
CB609032.1 - 3, 4 549

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
histone H3 acetylation
molecular_function
identical protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0