This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other AGMO SIRNA Assays ›
Gene Symbol:
Agmo
Gene Name:
alkylglycerol monooxygenase
Gene Aliases:
RGD1312038, Tmem195
Chromosome Location:
Chr.6: 56846859 - 57193918 on Build Rnor_6.0
UniGene ID:
Rn.152690
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s169657

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Agmo
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135899.1 NP_001129371.1 1 192
GenBank mRNA BC127545.1 - 1 192 7321962
FQ211046.1 - 1 248
FQ219104.1 - 1 236

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_178767, AK040702, AK039191, AK050246, AK163155.

Gene Ontology Categories:

Function(s) Process(es)

fatty acid biosynthetic process
membrane lipid metabolic process
ether lipid metabolic process
oxidation-reduction process
iron ion binding
glyceryl-ether monooxygenase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0