This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PPP1R21 SIRNA Assays ›
Gene Symbol:
Ppp1r21
Gene Name:
protein phosphatase 1, regulatory subunit 21
Gene Aliases:
RGD1565310
Chromosome Location:
Chr.6: 12253788 - 12323421 on Build Rnor_6.0
UniGene ID:
Rn.14962
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s169589

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ppp1r21
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271305.1 NP_001258234.1 9 1002
XM_008764482.1 XP_008762704.1 8 915
GenBank mRNA AY390384.1 - 9 853

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_028658, AK155094, AK170254, AK182861, BC137673, and others.

Gene Ontology Categories:

Process(es)

phosphatase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0