This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RCAN3 SIRNA Assays ›
Gene Symbol:
Rcan3
Gene Name:
RCAN family member 3
Gene Aliases:
Dscr1l2
Chromosome Location:
Chr.5: 153773461 - 153790272 on Build Rnor_6.0
UniGene ID:
Rn.162116
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s169480

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rcan3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012746.1 NP_001012764.1 4 712
XM_006239218.2 XP_006239280.1 5 931
GenBank mRNA AY919305.1 - 4 712

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_022980, AK043916, BC059001, AF237888, AK042201, and others.

Gene Ontology Categories:

Function(s) Process(es)

calcium-mediated signaling
troponin I binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0