This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GNL2 SIRNA Assays ›
Gene Symbol:
Gnl2
Gene Name:
guanine nucleotide binding protein-like 2 (nucleolar)
Gene Aliases:
RGD1305006
Chromosome Location:
Chr.5: 143028031 - 143053418 on Build Rnor_6.0
UniGene ID:
Rn.18964
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s169407

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gnl2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025736.1 NP_001020907.1 4 418
GenBank mRNA BC099173.1 - 4 418 7382617

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_145552, AK037796, BC003262, AK077532, AK146404, and others.

Gene Ontology Categories:

Function(s) Process(es)

metabolic process
ribosome biogenesis
GTPase activity
GTP binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0