This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FGFR1OP2 SIRNA Assays ›
Gene Symbol:
Fgfr1op2
Gene Name:
FGFR1 oncogene partner 2
Gene Aliases:
Wit3.0
Chromosome Location:
Chr.4: 180887182 - 180908206 on Build Rnor_6.0
UniGene ID:
Rn.68658
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s169176

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fgfr1op2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_201421.1 NP_958824.1 2 404
XM_006237688.2 XP_006237750.1 2 393
XM_008763402.1 XP_008761624.1 2 383
GenBank mRNA AY426739.1 - 2 404
AY426740.1 - 2 404
BC087696.1 - 2 301 7312267

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

response to wounding
wound healing
molecular_function
protein homodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0