siRNA Details
Gene Symbol: Il17re | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004091.1 | NP_001004091.1 | 15 | 1869 | ||
GenBank mRNA | BC061739.1 | - | 3 | 586 | ||
BC078742.1 | - | 15 | 1869 | 7123764 |

Gene Symbol: Il17re | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004091.1 | NP_001004091.1 | 15 | 1869 | ||
GenBank mRNA | BC061739.1 | - | 3 | 586 | ||
BC078742.1 | - | 15 | 1869 | 7123764 |