This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other AKAP4 SIRNA Assays ›
Gene Symbol:
AKAP4
Gene Name:
A-kinase anchoring protein 4
Gene Aliases:
AKAP 82, AKAP-4, AKAP82, CT99, FSC1, HI, PRKA4, hAKAP82, p82
Chromosome Location:
Chr.X: 50190768 - 50201013 on Build GRCh38
UniGene ID:
Hs.97633
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s16901

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: AKAP4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003886.2 NP_003877.2 2 207
NM_139289.1 NP_647450.1 2 180
GenBank mRNA AF072756.1 - 2 235
AK292463.1 - 2 201
BC033007.1 - 2 204
BC070150.1 - 2 199
BC126250.1 - 2 206 8991966
BC126252.1 - 2 206 8991968
Y15195.1 - 2 207

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

movement of cell or subcellular component
signal transduction
transmembrane receptor protein serine/threonine kinase signaling pathway
single fertilization
regulation of protein kinase A signaling
sperm motility
motile cilium assembly
establishment of protein localization
protein kinase A binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0