This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LANCL2 SIRNA Assays ›
Gene Symbol:
Lancl2
Gene Name:
LanC lantibiotic synthetase component C-like 2 (bacterial)
Gene Aliases:
-
Chromosome Location:
Chr.4: 88524212 - 88565113 on Build Rnor_6.0
UniGene ID:
Rn.104463
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s169008

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Lancl2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014187.1 NP_001014209.1 6 937
XM_006236580.2 XP_006236642.2 6 799
XM_006236581.2 XP_006236643.1 6 946
GenBank mRNA BC079412.1 - 6 937 7113320

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

metabolic process
positive regulation of abscisic acid-activated signaling pathway
negative regulation of transcription, DNA-templated
catalytic activity
phosphatidylinositol-5-phosphate binding
phosphatidylinositol-3-phosphate binding
phosphatidylinositol-4-phosphate binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0