This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TFAP2C SIRNA Assays ›
Gene Symbol:
Tfap2c
Gene Name:
transcription factor AP-2 gamma
Gene Aliases:
AP2-gamma, Tcfap2c
Chromosome Location:
Chr.3: 170550314 - 170559202 on Build Rnor_6.0
UniGene ID:
Rn.16142
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s168842

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tfap2c
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_201420.2 NP_958823.2 7 1398
XM_006235691.2 XP_006235753.1 7 1367
GenBank mRNA AY510602.1 - 7 1189
BC097260.1 - 7 1398 7443035

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_009335, NM_001159696, X94694, BC003778, AK145166, and others.

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
trophectodermal cell differentiation
hair follicle development
keratinocyte development
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
biological_process
male gonad development
stem cell population maintenance
forebrain neuron fate commitment
cerebral cortex development
cell differentiation
germ-line stem cell population maintenance
somatic stem cell population maintenance
regulation of gene expression, epigenetic
regulation of cell proliferation
skin development
regulation of epidermis development
positive regulation of transcription from RNA polymerase II promoter
sebaceous gland development
anatomical structure development
dichotomous subdivision of terminal units involved in mammary gland duct morphogenesis
epithelial cell proliferation involved in mammary gland duct elongation
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
core promoter binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
molecular_function
DNA binding
transcription factor activity, sequence-specific DNA binding
protein dimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0