This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PROCR SIRNA Assays ›
Gene Symbol:
Procr
Gene Name:
protein C receptor, endothelial
Gene Aliases:
-
Chromosome Location:
Chr.3: 151285321 - 151289581 on Build Rnor_6.0
UniGene ID:
Rn.144312
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s168780

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Procr
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025733.2 NP_001020904.1 3 657
GenBank mRNA BC097380.1 - 3 628 7445753
BC099115.1 - 3 657 7462042
FQ220150.1 - 3 696
FQ221148.1 - 3 664
FQ228484.1 - 3 662
FQ228710.1 - 3 664
FQ229445.1 - 3 694
FQ229483.1 - 3 664

Back To Top

More Information


Gene Ontology Categories:

Function(s)

immune response
blood coagulation
antigen processing and presentation
negative regulation of coagulation

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0