This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other UBAC1 SIRNA Assays ›
Gene Symbol:
Ubac1
Gene Name:
UBA domain containing 1
Gene Aliases:
Ubadc1
Chromosome Location:
Chr.3: 3453396 - 3476242 on Build Rnor_6.0
UniGene ID:
Rn.6512
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s168493

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ubac1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007742.1 NP_001007743.1 4, 5 601
XM_006233689.1 XP_006233751.1 4, 5 628
GenBank mRNA BC083603.1 - 4, 5 601 7133164
FQ210767.1 - 4, 5 487

Back To Top

More Information


Gene Ontology Categories:

Function(s)

protein ubiquitination

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0