siRNA Details
Gene Symbol: RGD1309170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134579.1 | NP_001128051.1 | 5, 6 | 945 | ||
XM_006233387.2 | XP_006233449.1 | 6, 7 | 841 |

Gene Symbol: RGD1309170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134579.1 | NP_001128051.1 | 5, 6 | 945 | ||
XM_006233387.2 | XP_006233449.1 | 6, 7 | 841 |