This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LOC103690070 SIRNA Assays ›
Gene Symbol:
LOC103690070 Cxadrl1
Gene Name:
coxsackievirus and adenovirus receptor-like
coxsackie virus and adenovirus receptor-like 1
Gene Aliases:
-
-
Chromosome Location:
Chr.8: 106832986 - 106859328 on Build Rnor_6.0
Chromosome Location:
Chr.8: 107246656 - 107272996 on Build Rnor_6.0
UniGene ID:
-
Rn.204925
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID s168157

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LOC103690070
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008766579.1 XP_008764801.1 1154
XM_008766580.1 XP_008764802.1 1154
XM_008766581.1 XP_008764803.1 474
Gene Symbol: Cxadrl1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199406.2 NP_955438.1 3 367
XM_008766539.1 XP_008764761.1 3 1154
XM_008766540.1 XP_008764762.1 2 474
GenBank mRNA BC063181.1 - 3 372 6887452

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0