This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NFKBIL1 SIRNA Assays ›
Gene Symbol:
Nfkbil1
Gene Name:
nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor-like 1
Gene Aliases:
-
Chromosome Location:
Chr.20: 4823612 - 4839202 on Build Rnor_6.0
UniGene ID:
Rn.38632
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s168016

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Nfkbil1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_212509.2 NP_997674.1 2, 3 440
XM_006256046.2 XP_006256108.1 2, 3 375
GenBank mRNA BC099772.1 - 2, 3 384 7932537

Back To Top

More Information


Gene Ontology Categories:

Function(s)

I-kappaB kinase/NF-kappaB signaling
negative regulation of lipopolysaccharide-mediated signaling pathway
negative regulation of NF-kappaB transcription factor activity
negative regulation of tumor necrosis factor production
negative regulation of toll-like receptor signaling pathway
cellular response to lipopolysaccharide

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header