This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TMEM138 SIRNA Assays ›
Gene Symbol:
Tmem138
Gene Name:
transmembrane protein 138
Gene Aliases:
-
Chromosome Location:
Chr.1: 226623861 - 226630933 on Build Rnor_6.0
UniGene ID:
Rn.102785
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s167895

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tmem138
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198777.2 NP_942072.2 4 518
XM_006231046.1 XP_006231108.1 4 611
XM_006231047.1 XP_006231109.1 4 645
XM_006231048.2 XP_006231110.1 4 400
GenBank mRNA BC061578.1 - 4 448
BC161808.1 - 4 790 9035805
FQ220672.1 - 4 523

Back To Top

More Information


Gene Ontology Categories:

Function(s)

cilium assembly

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0