This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TOLLIP SIRNA Assays ›
Gene Symbol:
Tollip
Gene Name:
toll interacting protein
Gene Aliases:
-
Chromosome Location:
Chr.1: 214823993 - 214844861 on Build Rnor_6.0
UniGene ID:
Rn.137040
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s167786

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tollip
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109668.1 NP_001103138.1 3 504
XM_006230607.2 XP_006230669.1 4 585
XM_008760003.1 XP_008758225.1 3 449
GenBank mRNA BC133067.1 - 3 390 7193007

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

autophagy
inflammatory response
signal transduction
phosphorylation
epithelial cell differentiation
positive regulation of protein sumoylation
protein localization to endosome
innate immune response
interleukin-1, Type I receptor binding
kinase binding
ubiquitin conjugating enzyme binding
ubiquitin protein ligase binding
SUMO binding
Toll-like receptor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-fd98957d6-rbs6c:80/100.66.128.142:80.
git-commit: 278a916a254c4ca07f9ccbb2b9efa8cd7578b5be
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0