This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TM6SF1 SIRNA Assays ›
Gene Symbol:
Tm6sf1
Gene Name:
transmembrane 6 superfamily member 1
Gene Aliases:
-
Chromosome Location:
Chr.1: 143657607 - 143703829 on Build Rnor_6.0
UniGene ID:
Rn.17383
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s167618

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tm6sf1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108490.1 NP_001101960.1 6, 7 610
XM_006229441.2 XP_006229503.1 5, 6 492
XM_008759543.1 XP_008757765.1 4, 5 872
XM_008759544.1 XP_008757766.1 5, 6 464

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0