This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RBM42 SIRNA Assays ›
Gene Symbol:
Rbm42
Gene Name:
RNA binding motif protein 42
Gene Aliases:
RGD1306184
Chromosome Location:
Chr.1: 89099296 - 89109251 on Build Rnor_6.0
UniGene ID:
Rn.17083
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s167531

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rbm42
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014159.2 NP_001014181.2 9 1263
XM_006228781.2 XP_006228843.1 8 1201
GenBank mRNA BC079321.1 - 9 1221 7135399
FQ216075.1 - 5 645
FQ222852.1 - 8 1146
FQ235070.1 - 5 613

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_024321, BC031682, BC002868, BC004204.
This siRNA also targets the following transcripts in Mouse: NM_133693, AK151000, BY551742, AK189364, AK215240, and others.

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of mRNA splicing, via spliceosome
nucleotide binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0