siRNA Details
Gene Symbol: Prodh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038588.1 | NP_001033677.1 | 5 | 638 | ||
GenBank mRNA | AF222852.1 | - | 5 | 628 | ||
BC158806.1 | - | 5 | 638 | 7324442 |

Gene Symbol: Prodh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038588.1 | NP_001033677.1 | 5 | 638 | ||
GenBank mRNA | AF222852.1 | - | 5 | 628 | ||
BC158806.1 | - | 5 | 638 | 7324442 |