This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PRMT7 SIRNA Assays ›
Gene Symbol:
Prmt7
Gene Name:
protein arginine methyltransferase 7
Gene Aliases:
RGD1304869
Chromosome Location:
Chr.19: 38189605 - 38237155 on Build Rnor_6.0
UniGene ID:
Rn.14954
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s167294

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Prmt7
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014153.1 NP_001014175.1 4 381
GenBank mRNA BC085121.1 - 4 381 7115238
FQ211965.1 - 424
FQ212485.1 - 348

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_145404, BC057177, BC012845, AK154255, AK173304, and others.

Gene Ontology Categories:

Function(s) Process(es)

spliceosomal snRNP assembly
regulation of gene expression by genetic imprinting
transcription, DNA-templated
regulation of transcription, DNA-templated
biological_process
histone methylation
peptidyl-arginine methylation
peptidyl-arginine methylation, to symmetrical-dimethyl arginine
peptidyl-arginine methylation, to asymmetrical-dimethyl arginine
cell differentiation
histone arginine methylation
DNA methylation involved in gamete generation
histone H4-R3 methylation
molecular_function
histone-arginine N-methyltransferase activity
S-adenosylmethionine-dependent methyltransferase activity
[myelin basic protein]-arginine N-methyltransferase activity
protein-arginine omega-N monomethyltransferase activity
protein-arginine omega-N asymmetric methyltransferase activity
protein-arginine omega-N symmetric methyltransferase activity
ribonucleoprotein complex binding
histone methyltransferase activity (H4-R3 specific)

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header