siRNA Details
Gene Symbol: Mocos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108425.1 | NP_001101895.1 | 9 | 1827 | ||
XM_006254466.2 | XP_006254528.1 | 8 | 1994 |

Gene Symbol: Mocos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108425.1 | NP_001101895.1 | 9 | 1827 | ||
XM_006254466.2 | XP_006254528.1 | 8 | 1994 |