This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RNF44 SIRNA Assays ›
Gene Symbol:
Rnf44
Gene Name:
ring finger protein 44
Gene Aliases:
-
Chromosome Location:
Chr.17: 10458029 - 10472410 on Build Rnor_6.0
UniGene ID:
Rn.21345
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s166982

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rnf44
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024795.1 NP_001019966.1 7 1110
XM_006253633.2 XP_006253695.1 7 2107
XM_006253634.2 XP_006253696.1 7 1100
XM_006253637.2 XP_006253699.1 7 2629
XM_006253641.2 XP_006253703.1 7 2043
XM_006253642.2 XP_006253704.1 7 2565
XM_006253644.2 XP_006253706.1 7 1036
XM_008771498.1 XP_008769720.1 7 2626
GenBank mRNA BC098030.1 - 7 1110 7456296

Back To Top

More Information


Gene Ontology Categories:

Process(es)

zinc ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0