This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DMTN SIRNA Assays ›
Gene Symbol:
Dmtn
Gene Name:
dematin actin binding protein
Gene Aliases:
Epb4.9, Epb49
Chromosome Location:
Chr.15: 52292762 - 52320385 on Build Rnor_6.0
UniGene ID:
Rn.9618
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s166774

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dmtn
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108385.1 NP_001101855.1 10 1117
XM_006252279.2 XP_006252341.1 9 1252
XM_006252280.2 XP_006252342.1 9 1250
XM_006252281.1 XP_006252343.1 7 605
XM_008770830.1 XP_008769052.1 10 1115
XM_008770831.1 XP_008769053.1 10 1772
GenBank mRNA BC166453.1 - 9 1126 8374835

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein complex assembly
regulation of cell shape
regulation of lamellipodium assembly
positive regulation of fibroblast migration
negative regulation of peptidyl-threonine phosphorylation
negative regulation of cell-substrate adhesion
actin cytoskeleton organization
positive regulation of blood coagulation
regulation of actin cytoskeleton organization
negative regulation of peptidyl-serine phosphorylation
calcium-mediated signaling using intracellular calcium source
calcium-mediated signaling using extracellular calcium source
erythrocyte development
negative regulation of peptidyl-tyrosine phosphorylation
actin filament bundle assembly
regulation of filopodium assembly
negative regulation of focal adhesion assembly
protein secretion by platelet
cellular response to calcium ion
cellular response to cAMP
positive regulation of wound healing
negative regulation of protein targeting to membrane
actin filament reorganization
negative regulation of substrate adhesion-dependent cell spreading
positive regulation of substrate adhesion-dependent cell spreading
positive regulation of platelet aggregation
positive regulation of integrin-mediated signaling pathway
actin binding
receptor binding
spectrin binding
protein self-association

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0