siRNA Details
Gene Symbol: Bin3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013186.1 | NP_001013204.1 | 6 | 356 | ||
GenBank mRNA | BC079146.1 | - | 6 | 356 | 7128352 |

Gene Symbol: Bin3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013186.1 | NP_001013204.1 | 6 | 356 | ||
GenBank mRNA | BC079146.1 | - | 6 | 356 | 7128352 |