siRNA Details
Gene Symbol: Tbc1d10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015022.3 | NP_001015022.1 | 3 | 385 | ||
GenBank mRNA | AB210856.1 | - | 3 | 385 | ||
BC093603.1 | - | 3 | 448 | 7316341 |

Gene Symbol: Tbc1d10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015022.3 | NP_001015022.1 | 3 | 385 | ||
GenBank mRNA | AB210856.1 | - | 3 | 385 | ||
BC093603.1 | - | 3 | 448 | 7316341 |