This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RUFY3 SIRNA Assays ›
Gene Symbol:
Rufy3
Gene Name:
RUN and FYVE domain containing 3
Gene Aliases:
Ripx, Singar1
Chromosome Location:
Chr.14: 21058575 - 21144743 on Build Rnor_6.0
UniGene ID:
Rn.228806
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s166540

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rufy3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025127.1 NP_001020298.1 4 897
XM_006250781.2 XP_006250843.1 4 1761
XM_006250782.2 XP_006250844.1 4 1970
XM_006250783.2 XP_006250845.1 4 1760
XM_006250784.2 XP_006250846.1 3 486
XM_006250785.2 XP_006250847.1 3 680
XM_008770044.1 XP_008768266.1 3 698
XM_008770045.1 XP_008768267.1 3 698
XM_008770046.1 XP_008768268.1 4 1955
XM_008770047.1 XP_008768269.1 3 698
XM_008770048.1 XP_008768270.1 5 1814
GenBank mRNA BC089952.1 - 4 897 7312240
EF538802.1 - 4 502
EF577045.1 - 3 448

Back To Top

More Information


Gene Ontology Categories:

Function(s)

regulation of axonogenesis
negative regulation of axonogenesis
positive regulation of axonogenesis

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0