This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other BRCA2 SIRNA Assays ›
Gene Symbol:
Brca2
Gene Name:
breast cancer 2, early onset
Gene Aliases:
-
Chromosome Location:
Chr.12: 504007 - 544754 on Build Rnor_6.0
UniGene ID:
Rn.103225
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s165603

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Brca2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031542.2 NP_113730.2 11 3949
GenBank mRNA U89653.1 - 11 3948

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

double-strand break repair via homologous recombination
oocyte maturation
inner cell mass cell proliferation
nucleotide-excision repair
double-strand break repair
DNA recombination
regulation of transcription, DNA-templated
cellular response to DNA damage stimulus
DNA damage response, signal transduction by p53 class mediator resulting in transcription of p21 class mediator
male meiosis I
spermatogenesis
brain development
cell aging
response to nutrient
cell proliferation
female gonad development
intrinsic apoptotic signaling pathway in response to DNA damage
response to X-ray
response to UV-C
response to gamma radiation
hemopoiesis
mammary gland development
homologous chromosome orientation involved in meiotic metaphase I plate congression
response to estradiol
regulation of cytokinesis
negative regulation of mammary gland epithelial cell proliferation
multicellular organism growth
regulation of cell proliferation
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
chordate embryonic development
histone H3 acetylation
histone H4 acetylation
positive regulation of transcription, DNA-templated
positive regulation of mitotic cell cycle
replication fork protection
chromosome organization
centrosome duplication
protease binding
single-stranded DNA binding
histone acetyltransferase activity
protein binding
H3 histone acetyltransferase activity
H4 histone acetyltransferase activity
gamma-tubulin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0