This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TBX3 SIRNA Assays ›
Gene Symbol:
Tbx3
Gene Name:
T-box 3
Gene Aliases:
-
Chromosome Location:
Chr.12: 42479518 - 42495167 on Build Rnor_6.0
UniGene ID:
Rn.162144
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s165547

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tbx3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181638.1 NP_853669.1 2 480
XM_006249415.2 XP_006249477.1 3 1628
XM_006249416.2 XP_006249478.1 3 2491
XM_006249417.2 XP_006249479.1 3 1638
XM_006249418.1 XP_006249480.1 2 1752
GenBank mRNA AY289619.1 - 2 480
FQ221880.1 - 2 1450

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
skeletal system development
blood vessel development
in utero embryonic development
heart looping
heart morphogenesis
outflow tract morphogenesis
atrioventricular bundle cell differentiation
transcription, DNA-templated
cell aging
positive regulation of cell proliferation
anterior/posterior axis specification, embryo
organ morphogenesis
specification of organ position
stem cell population maintenance
limbic system development
male genitalia development
female genitalia development
negative regulation of epithelial cell differentiation
mammary gland development
luteinizing hormone secretion
embryonic heart tube development
limb morphogenesis
embryonic forelimb morphogenesis
embryonic hindlimb morphogenesis
forelimb morphogenesis
regulation of cell proliferation
embryonic digit morphogenesis
negative regulation of apoptotic process
negative regulation of myoblast differentiation
positive regulation of cell cycle
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
follicle-stimulating hormone secretion
mesoderm morphogenesis
cardiac muscle cell differentiation
palate development
ventricular septum morphogenesis
branching involved in mammary gland duct morphogenesis
mammary placode formation
cardiac muscle cell fate commitment
sinoatrial node cell development
cellular senescence
positive regulation of stem cell proliferation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II transcription factor binding
RNA polymerase II activating transcription factor binding
molecular_function
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0