This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZDHHC15 SIRNA Assays ›
Gene Symbol:
Zdhhc15
Gene Name:
zinc finger, DHHC-type containing 15
Gene Aliases:
-
Chromosome Location:
Chr.X: 75433961 - 75566538 on Build Rnor_6.0
UniGene ID:
Rn.214711
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s165142

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Zdhhc15
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039101.1 NP_001034190.1 7 739
XM_006257145.2 XP_006257207.1 7 796
XM_008773347.1 XP_008771569.1 8 1559
XM_008773348.1 XP_008771570.1 9 712
GenBank mRNA AY886531.1 - 7 739

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

synaptic vesicle maturation
protein palmitoylation
establishment of protein localization
zinc ion binding
palmitoyltransferase activity
protein-cysteine S-palmitoyltransferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header