This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CCDC150 SIRNA Assays ›
Gene Symbol:
Ccdc150
Gene Name:
coiled-coil domain containing 150
Gene Aliases:
RGD1304775
Chromosome Location:
Chr.9: 60883969 - 60988898 on Build Rnor_6.0
UniGene ID:
Rn.214105
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s164818

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ccdc150
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191806.1 NP_001178735.1 21 2537
XM_006244924.2 XP_006244986.1 21 2693
XM_008767094.1 XP_008765316.1 22 2759
XM_008767095.1 XP_008765317.1 21 2693
XM_008767096.1 XP_008765318.1 21 2693
XM_008767097.1 XP_008765319.1 21 2693

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0