This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NBEAL2 SIRNA Assays ›
Gene Symbol:
Nbeal2
Gene Name:
neurobeachin-like 2
Gene Aliases:
Nbeal2-ps1, RGD1306501
Chromosome Location:
Chr.8: 118893987 - 118924392 on Build Rnor_6.0
UniGene ID:
-
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s164530

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Nbeal2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003750590.3 XP_003750638.1 34 5774
XM_003750591.3 XP_003750639.1 33 5693
XM_006244008.2 XP_006244070.1 34 7001
XM_006244009.2 XP_006244071.1 34 5775

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

blood coagulation
endosomal transport
platelet formation
megakaryocyte development
wound healing
platelet alpha granule organization
phospholipid binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0