This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RNF128 SIRNA Assays ›
Gene Symbol:
Rnf128
Gene Name:
ring finger protein 128, E3 ubiquitin protein ligase
Gene Aliases:
RGD1566282
Chromosome Location:
Chr.X: 110789057 - 110877964 on Build Rnor_6.0
UniGene ID:
Rn.7002
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s164401

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rnf128
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008773419.1 XP_008771641.1 1 698
GenBank mRNA BC160840.1 - 1 486 7460350

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein ubiquitination
negative regulation of cytokine biosynthetic process
ubiquitin-protein transferase activity
zinc ion binding
ubiquitin protein ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0