This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SENP8 SIRNA Assays ›
Gene Symbol:
Senp8
Gene Name:
SUMO/sentrin specific peptidase family member 8
Gene Aliases:
-
Chromosome Location:
Chr.8: 64558506 - 64572964 on Build Rnor_6.0
UniGene ID:
Rn.145873
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s164253

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Senp8
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012355.1 NP_001012355.1 3 523
XM_006243196.2 XP_006243258.1 2 617
GenBank mRNA BC089939.1 - 3 523 7310036
FQ226292.1 - 2 505
FQ227853.1 - 3 536
FQ231841.1 - 2 515

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein deneddylation
proteolysis
NEDD8-specific protease activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-6889b944d9-flx6z:80/100.66.128.142:80.
git-commit: df865406a845caddbd23a531a4b79e504db49c75
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header