This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CDK11B SIRNA Assays ›
Gene Symbol:
Cdk11b
Gene Name:
cyclin-dependent kinase 11B
Gene Aliases:
Cdc2l1, Cdk11
Chromosome Location:
Chr.5: 173051898 - 173078049 on Build Rnor_6.0
UniGene ID:
Rn.10204
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s163952

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cdk11b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145766.2 NP_665709.2 7 712
XM_006239527.2 XP_006239589.1 6 1268
XM_006239528.2 XP_006239590.1 7 1075
XM_006239529.2 XP_006239591.1 7 996
XM_006239530.1 XP_006239592.1 7 696
XM_006239531.2 XP_006239593.1 7 800
XM_006239532.2 XP_006239594.1 2 1144
XM_006239533.1 XP_006239595.1 2 121
GenBank mRNA DY556124.1 - 2 132

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of cell growth
blastocyst development
protein phosphorylation
regulation of mitotic nuclear division
regulation of mRNA processing
regulation of cell cycle
negative regulation of apoptotic signaling pathway
protein serine/threonine kinase activity
cyclin-dependent protein serine/threonine kinase activity
ATP binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0