This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZFP707 SIRNA Assays ›
Gene Symbol:
Zfp707 RGD1311188
Gene Name:
zinc finger protein 707
similar to 1500031N24Rik protein
Gene Aliases:
RGD1562665, Zfp979, Znf707
-
Chromosome Location:
Chr.7: 116996615 - 117007663 on Build Rnor_6.0
Chromosome Location:
Chr.7: 117025917 - 117032875 on Build Rnor_6.0
UniGene ID:
Rn.152841
Rn.224779
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID s163680

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Zfp707
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135903.1 NP_001129375.1 2, 3 404
XM_006241815.2 XP_006241877.1 2, 3 634
XM_006241816.2 XP_006241878.1 2, 3 632
GenBank mRNA BC161959.1 - 2, 3 404 9027463
Gene Symbol: RGD1311188
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135897.1 NP_001129369.1 2, 3 410
XM_008765570.1 XP_008763792.1 2, 3 381
GenBank mRNA BC162017.1 - 2, 3 410 9098233

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001081065, BC026404, AK137962, AK153958, AK005330.

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
nucleic acid binding
transcription factor activity, sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header